Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Lugol's iodine solution reagent B for PREVI® COLOR GRAM instrument
Supplier: bioMérieux
Danger
29586 29586
89127-614EA 38.85 USD InStock
CA-89127-614 89127-614
Lugol's iodine solution reagent B for PREVI® COLOR GRAM instrument
Lugol's iodine solution
RB IODINE solution
  • Easy to use
  • Premade reagents
  • No mess in the lab
The PREVI® Color Gram dyes are used to stain micro-organisms according to Gram’s method. They are intended for use exclusively with the PREVI® Color Gram instrument.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty