Would you like to log in again?
ViroGuard® Blue Coveralls with Hood, Elastic Wrist and Back, Front Zipper with Storm Flap, International Enviroguard™
Supplier: International Enviroguard
2407-XL 2407-2XL 2407-L 2407-3XL 2407-5XL 2407-M
89127-684CS 509.25 USD InStock
89127-684 89127-682 89127-678 89127-688 89127-676 89127-680
ViroGuard® Blue Coveralls with Hood, Elastic Wrist and Back, Front Zipper with Storm Flap, International Enviroguard™
Disposable Garments
Disposable Coverall with fabric that has been tested for protection against blood, blood-borne pathogens, and many other liquid hazards.
  • Strong fabric provides excellent abrasion resistance and resists tears
  • Helps to keep the wearer cool and avoid heat stress through enhanced moisture vapor transmission
  • Attached hood with elastic wrists and ankles for snug, secure fit
  • Zipper front with sealable storm flap for an additional layer of protection
  • Blue color makes stains and splashes on fabric less conspicuous
Fabric passes ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and blood-borne pathogens. Roomy, seamless shoulder area resists tears and provides more protection in key exposure area. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with user for more comfort and better tear resistance.
Recommended applications include emergency response, category 3 black water, tank cleaning, crime scene remediation, and animal research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty