Would you like to log in again?
MicroGuard CE™ Coveralls, International ENVIROGUARD™
Supplier: International Enviroguard
MicroGuard CE™
CE8013CI-M CE8019CI-M CE8019CI-3XL CE8013CI-XL CE8013CI-3XL CE8013CI-L CE8019CI-4XL CE8019CI-2XL CE8019CI-L CE8013CI-2XL CE8013CI-4XL CE8019CI-XL
89127-710CS 475.07 USD InStock
89127-710 89127-732 89127-730 89127-726 89127-704 89127-736 89127-702 89127-734 89127-712 89127-708 89127-706 89127-728
MicroGuard CE™ Coveralls, International ENVIROGUARD™
Disposable Garments
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting
Coveralls are white with tunnelized wrists and ankles.
Tear-resistant. Recyclable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty