Would you like to log in again?
MicroGuard CE™ Frocks, International ENVIROGUARD™
Supplier: International Enviroguard
MicroGuard CE™
CE8046CI-L CE8046CI-3XL CE8046CI-XL CE8046CI-4XL CE8046CI-M CE8046CI-2XL
89127-784CS 569.47 USD InStock
89127-784 89127-782 89127-776 89127-774 89127-778 89127-780
MicroGuard CE™ Frocks, International ENVIROGUARD™
Disposable Garments
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting.
Frocks are white with no pockets and tunnelized wrists. Tear-resistant. Recyclable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty