Would you like to log in again?
MicroGuard CE™ Boot Covers, International ENVIROGUARD™
Supplier: International Enviroguard
MicroGuard CE™
CE8104CI-L CE8106CI-XL
89127-808CS 772.61 USD InStock
89127-808 89127-804
MicroGuard CE™ Boot Covers, International ENVIROGUARD™
Disposable Garments
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting
Tear-resistant. Recyclable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty