Would you like to log in again?
GammaGuard CE™ Shoe Covers, Sterile, International ENVIROGUARD™
Supplier: International Enviroguard
GammaGuard CE™
CE11101CIS-L
89127-972CS 828.53 USD InStock
89127-972
GammaGuard CE™ Shoe Covers, Sterile, International ENVIROGUARD™
Disposable Garments
GammaGuard CE™ sterile cleanroom apparel offers a superior liquid barrier, breathability, low linting, and tear resistance
Shoe covers have non-skid soles. Shoe covers are sterilized to a 10-6 Sterility Assurance Level (SAL).
Special fold design prevents contamination during gowning. Material contains no natural rubber latex. Recyclable.
Ordering information: Manufactured in an ISO-certified facility, clean processed, and individually packaged under cleanroom conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty