Would you like to log in again?
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Supplier: Argos Technologies
PolarSafe™
04396-55 04396-56
89128-210CS 441.39 USD InStock
89128-210 89128-208
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Boxes Cryoboxes
Polypropylene freezer boxes accommodate 15 & 50mL centrifuge tubes for long term storage; an excellent alternative to cardboard storage boxes
Boxes can be stored to -90°C (130°F) and are autoclavable to 122°C (251.6°F). Accommodate most 4" stainless steel freezer racks.
Clear lid and easy-to-read numerical grid allows for tube reference and identification. White top portion allows for easy labeling or marking.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty