Would you like to log in again?
PolarSafe™ Polycarbonate Freezer Storage Boxes, Argos Technologies
Supplier: Argos Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PolarSafe™
04398-04 04398-05 04398-06 04398-07
89128-212CS 842.32 USD InStock
89128-212 89128-214 89128-216 89128-218
PolarSafe™ Polycarbonate Freezer Storage Boxes, Argos Technologies
Boxes Cryoboxes
Polycarbonate freezer boxes, available with 25, 81 and 100-place divisions
Boxes have clear lids with locator grids to facilitate sample identification. Boxes are designed to withstand temperatures from -196° to 121° C. Boxes have drain holes for venting and safety. Keyed lid prevents misalignment of grid. Tall box (51/4 x 51/4 x 33/4 ") accommodates tall tubes & vials (4-5mL).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty