Would you like to log in again?
VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Signature™
IC181SWH3X0025VD IC181SWHSM0025VD IC181SWHSM0025VD IC190SWH2XVD IC181SWHXL0025VD IC190SWHMDVD IC190SWHLGVD IC181SWHLG0025VD IC106SWH3X0025VD IC106SWHMD0025VD IC181SWHMD0025VD IC181SWHXL0025VD IC190SWHXLVD IC181SWH2X0025VD IC181SWH2X0025VD IC106SWH2X0025VD IC106SWHLG0025VD IC181SWH4X0025VD IC181SWHMD0025VD IC181SWH3X0025VD IC190SWH3XVD IC106SWHXL0025VD IC181SWHLG0025VD IC181SWH4X0025VD
89012-830CS 550.61 USD InStock
CA89012-830 89012-830 89012-822 89127-188 89012-832 80200-628 89012-824 CA89012-834 CA89012-824 89012-834 89012-826 CA89012-832 CA89012-822 89128-778 80200-622 80200-620 80200-626 80200-624 89128-780 89128-782 89128-784 89012-828 CA89012-828 CA89012-826
VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Protective, durable, and comfortable coveralls provide an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Protective, durable, and comfortable
  • Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Chemically and biologically inert, low linting, and static free until laundered
  • Serged seams, laydown collar, front zipper closure, and elastic wrists
  • Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood
  • Boot attachments combine embossed PVC exterior sole with textured interior to ensure secure, nonslip fit and feel
All coveralls feature serged seams, laydown collar, front zipper closure, and elastic wrists. Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood. Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, nonslip fit and feel.
Garments are chemically and biologically inert, low-linting, and static-free until laundered.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty