Would you like to log in again?
VWR® Signature™ Lab Coats made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Signature™
IC224SWHXL0030VD IC224SWHLG0030VD IC224SWH2X0030VD IC224SWHSM0030VD IC224SWHMD0030VD
89127-198CS 255.73 USD InStock
89127-198 89125-786 89128-798 89128-786 89127-192
VWR® Signature™ Lab Coats made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Garments provide a premium level of protection and durability while remaining comfortable
  • Premium level of protection and durability while remaining comfortable
  • Excellent barrier against dry particles, nonhazardous liquids, and microorganisms, designed for use in cleanroom environments
  • Limited-use lab coats are antistatic treated, low linting, chemically and biologically inert
Lab coats feature serged seams, laydown collar, five-snap front, one breast pocket, two hip pockets, and open wrists. All coats feature Raglan sleeves.
Lab coats offer an excellent barrier against dry particles, nonhazardous liquids, and microorganisms, and are designed for use in cleanroom environments. Limited-use lab coats are antistatic-treated, low-linting, and chemically and biologically inert.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty