Would you like to log in again?
VWR® Signature™ Frocks made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Signature™
IC264SWH2X0030VD IC270BWH3X0030VD IC270BWHMD0030VD IC270BWHSM0030VD IC264SWHSM0030VD IC264SWHLG0030VD IC270BWHXL0030VD IC270BWH2X0030VD IC270BWH4X0030VD IC270BWHLG0030VD IC264SWHXL0030VD IC264SWHMD0030VD
80200-608CS 719.73 USD InStock
80200-608 80200-606 89127-312 89127-308 80200-610 80200-600 89128-788 89125-608 89127-316 80200-604 80200-612 80200-602
VWR® Signature™ Frocks made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Frocks provide a high level of protection in cleanroom environments.
  • High level of protection in cleanroom environments
  • Chemically and biologically inert
  • Low linting
  • Static-free until laundered
These limited-use frocks are available in two styles. Frocks with covered zipper front feature tunneled elastic wrists and serged seams. These frocks are available with a bound neck. Frocks with five-snap front feature a bound neck and bound seams. All garments are pocketless.
Garments are chemically and biologically inert, low-linting, and static-free until laundered.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty