Would you like to log in again?
VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Signature™
IC451SWHLG0100VD IC451SWHXL0100VD IC451SWHMD0100VD
89128-792CS 239.27 USD InStock
89128-792 89128-794 89128-790
VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Shoe covers are manufactured using a unique flash spinning process that creates an excellent barrier against nonhazardous liquids and microorganisms as well as dry particles down to 0.5µm.
  • Excellent barrier against nonhazardous liquids and microorganisms, dry particles down to 0.5µm
  • For cleanroom use, chemically and biologically inert, tear resistant, and low linting
  • Single use, serged seams and elastic tops for a secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
Designed for cleanroom use, garments are chemically and biologically inert, tear-resistant, and low-linting. Single use shoe covers feature serged seams and elastic tops for a secure fit. Also equipped with flat, non-skid PVC soles.
Ordering information: Bulk packed.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty