Would you like to log in again?
Cy™ Amidites, Cytiva
Supplier: Cytiva
CY™
28-9172-98 28917298 28904250 28-9021-58 28-9021-60 28-9172-99 28-9042-50 28-9042-49 28904249 27-1798-01
95040-118EA 1070.88 USD InStock
95040-118 95040-126 89128-840 CA76541-098 CA76541-100 CA76541-102 89128-834 89128-836 95040-124 89128-838
Cy™ Amidites, Cytiva
Electrophoresis Reagents Western Blotting Reagents
Cy™ fluorescent amidites for incorporation of Cy3, Cy3,5, Cy5, Cy5,5 fluorescent dyes at any position in an oligonucleotide during synthesis.
  • For incorporation of Cy fluorescent dyes at any position in an oligonucleotide during synthesis
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
Cy5 amidite can be used to prepare sequencing primers for use on ALF express.
Delivery information: Larger pack sizes and custom packaging available upon request.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty