Would you like to log in again?
Disposable Columns Prepacked with Sepharose™, His SpinTrap™ Kit, Cytiva
Supplier: Cytiva
His SpinTrap™
28-9321-71 28-9321-71
89128-844EA 608.4 USD InStock
89128-844 CA89128-844
Disposable Columns Prepacked with Sepharose™, His SpinTrap™ Kit, Cytiva
Chromatography Columns
His SpinTrap is a prepacked, single use spin column for purifying histidine-tagged proteins by immobilised metal ion affinity chromatography (IMAC) and rapid expression screening.
  • Ni Sepharose high performance in prepacked spin columns
  • Yields up to 750 µg pure protein per column
  • Allows direct purification of both clarified and unclarified cell lysates
Delivery information: Kit contains one pack of His SpinTrap™ and one pack of His Buffer Kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty