Would you like to log in again?
SOURCE™ 15RPC Reversed Phase Chromatography Media, Cytiva
Supplier: Cytiva
SOURCE™
17-0727-20 17-0727-20
89128-850EA 846.16 USD InStock
89128-850 CA89128-850
SOURCE™ 15RPC Reversed Phase Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
SOURCE™ 15RPC is designed for reversed phase chromatography (RPC) with full pH range stability and suitable for polishing steps in industrial processes requiring high flow velocities and low back-pressure.
  • Mono-sized 15 µm rigid beads gives low back pressures and high flow rates allowing for high productivity
  • Designed for high resolution reversed phase chromatography of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • Bioprocess medium supported for industrial applications
A controlled pore size distribution and large specific surface area offer excellent capacity for a wide range of molecules, from small peptides and oligonucleotides up to large proteins. With full pH range stability, SOURCE™ 15RPC is a strong alternative to silica-based RPC matrices.
Available in a range of different bulk pack sizes for easy scale-up and process development.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty