Would you like to log in again?
SOURCE™ 30S Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
SOURCE™
17-1273-01 17-1273-01
89128-852EA 1105.87 USD InStock
89128-852 CA89128-852
SOURCE™ 30S Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
SOURCE 30S is a synthetic high performance, preparative, chromatography medium, based on a 30 µm monosized, rigid polystyrene/divinyl benzene polymer matrix It is modified with sulphonate (S) strong cation exchange groups.
  • Strong cation exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • A high degree of purification with high productivity
  • Bioprocessing medium supported for industrial applications
  • Particle structure is consistent both bead-to-bead and batch-to batch
SOURCE 30S is suitable for intermediate purification steps in industrial processes when large volumes of partially purified material need to be processed. A high degree of purification can be obtained with high productivity. Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. As member of the BioProcess media range, SOURCE 30S meets industrial demands with security of supply and comprehensive technical and regulatory support.
SOURCE media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance at both laboratory and process scales.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty