Would you like to log in again?
SOURCE™ 15Q Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
SOURCE™
17-0947-01 17-0947-20 17-0947-01 17-0947-20
89128-854EA 522.94 USD InStock
89128-854 89128-856 CA89128-854 CA89128-856
SOURCE™ 15Q Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Mono-sized 15 um rigid beads gives low back pressure and high flow rates allowing for high productivity.
  • Polymeric, strong anion exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • Bioprocessing medium supported for industrial applications
  • Particle structure is consistent both bead-to-bead and batch-to batch
SOURCE 15Q is a synthetic high performance, preparative, chromatography medium, based on a 15 µm monosized, rigid polystyrene/divinyl benzene polymer matrix. It is modified with quaternary ammonium (Q) strong anion exchange groups. SOURCE media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance.
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. With its very high resolution, SOURCE 15Q is suitable for polishing steps in in both research and industrial applications. As member of the BioProcess media range, SOURCE 15Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Strong anion exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty