Would you like to log in again?
SOURCE™ 15S Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SOURCE™
17-0944-10 17-0944-10 17-0944-01 17-0944-01
89128-860EA 504.88 USD InStock
89128-860 CA89128-860 89128-868 CA89128-868
SOURCE™ 15S Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
SOURCE 15S is a synthetic high performance, preparative, chromatography medium, based on a 15 µm monosized, rigid polystyrene/divinyl benzene polymer matrix.
  • Mono-sized 15 um rigid beads gives low back pressure and high flow rates allowing for high productivity
  • Sulphonate group, long, hydrophilic spacer arms coupling method
  • Strong cation exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • Stable in all common ion exchange buffers
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. As member of the BioProcess media range, SOURCE 15S meets industrial demands with security of supply and comprehensive technical and regulatory support.
It is modified with sulphonate (S) strong cation exchange groups. SOURCE media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance. With its very high resolution, SOURCE 15S is suitable for polishing steps in both research and industrial applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty