Would you like to log in again?
HiPrep SP XL 16/10 Columns, Cytiva
Supplier: Cytiva
HiPrep™
28-9365-40 28-9365-40
89128-882EA 932.23 USD InStock
89128-882 CA89128-882
HiPrep SP XL 16/10 Columns, Cytiva
Chromatography Columns
HiPrep SP XL 16/10 is a prepacked, ready to use column for ion exchange chromatography.
  • Provide high loading capacity and fast purifications
  • Optimized for reliable, reproducible separations
  • Compatible with simple pump-based configurations and chromatography systems such as ÄKTA design
  • Excellent choice for rapid enrichment during initial capture of proteins from start material
  • Strong Sulfopropyl (SP) cation exchanger
It is prepacked with SP Sepharose XL for inital capture of proteins and other biomoleculs with ion exchange (IEX) chromatography.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty