Would you like to log in again?
HiPrep CM FF Columns, Cytiva
Supplier: Cytiva
HiPrep™
28-9365-42 28-9365-42
89128-888EA 926.19 USD InStock
89128-888 CA89128-888
HiPrep CM FF Columns, Cytiva
Chromatography Columns
HiPrep CM FF 16/10 is a weak anion exchanger prepacked with CM Sepharose Fast Flow, ready-to-use for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography
  • 20 ml HiPrep columns provide fast, reliable, and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
  • Compatible with single-pump configurations and chromatography systems such as ÄKTA design
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty