Would you like to log in again?
HisPrep FF Columns, Cytiva
Supplier: Cytiva
HisPrep™
28-9365-51 28936551 28-9365-51
89128-894EA 659.72 USD InStock
CA95055-920L 89128-894 CA89128-894
HisPrep FF Columns, Cytiva
Chromatography Columns
HisPrep FF 16/10 is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow. The column is an excellent choice for preparative, scale-up purification of histidine-tagged recombinant proteins.
  • Optimized for convenient scale-up purification of histidine-tagged proteins
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Negligible leakage of Ni2+
  • High binding capacity, approx. 40 mg/ml resin
  • Convenient and time-saving 20 ml prepacked HiPrep format
HisPrep FF 16/10 columns are prepacked with Ni Sepharose 6 Fast Flow for scale-up purification of histidine-tagged proteins. Ni Sepharose 6 Fast Flow consists of 90 µm beads of highly cross-linked agarose, to which a chelating ligand has been immobilized.
This chelating ligand is charged with Ni2+ ions, the first-choice metal ion for purifying most histidine-tagged proteins. The negligible leakage of Ni2+ ions ensures reliable capture of histidine-tagged proteins in repeated IMAC purifications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty