Would you like to log in again?
GSTPrep FF 16/10 Columns, Cytiva
Supplier: Cytiva
GSTPrep™
28-9365-50 28-9365-50
89128-896EA 1047.3 USD InStock
89128-896 CA89128-896
GSTPrep FF 16/10 Columns, Cytiva
Chromatography Columns
GSTPrep FF 16/10 is prepacked, ready-to-use column containing Glutathione Sepharose 4 Fast Flow designed for one-step, scale-up purification of glutathione S-transferase (GST)-tagged proteins.
  • Binds approx. 200 mg recombinant GST/column
  • Mild elution conditions preserving protein antigenicity and function
  • Simple operation with a pump or chromatography system, such as ÄKTA design
Glutathione Sepharose 4 Fast Flow is for scale-up purification of recombinant GST-tagged proteins, other glutathione S-transferases, and glutathione-dependent proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty