Would you like to log in again?
Albumin and IgG Depletion SpinTrap, Cytiva
Supplier: Cytiva
SpinTrap™
28-9480-20 28-9480-20
89129-108PK 973.28 USD InStock
CA89129-108 89129-108
Albumin and IgG Depletion SpinTrap, Cytiva
Chromatography Columns
Albumin and IgG depletion SpinTrap™ columns for the depletion of albumin and IgG from human serum and plasma. Columns are prepacked with high performance Sepharose-based media with an affinity for human serum albumin (HSA) and IgG.
  • Handles smaller sample volumes (~50 μl) using a tabletop centrifuge
  • Columns are prepacked with high performance Sepharose based resin with an affinity for human serum albumin (HSA) and IgG
  • High depletion capacity, removes >95% HSA and >90% IgG, with high reproducibility
  • Simple and fast procedure
  • Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins
Albumin and IgG are the most abundant plasma proteins, and they tend to obscure the signals of less abundant proteins. Depletion of the two proteins removes more than 60% of the total protein content in human plasma, allowing a higher load of less abundant proteins to be included in the analysis and detection of more proteins. These columns are members of the Trap platform and address the need for flexible, small-scale preparation of protein samples prior to downstream analyses such as 1-D or 2-D gel electrophoresis and mass spectrometry.
For larger scale please use HiTrap Albumin and IgG Depletion columns.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty