Would you like to log in again?
DIGE Buffer Kit, Cytiva
Supplier: Cytiva
28-9374-52 28-9374-52
89129-110EA 356.92 USD InStock
CA89129-110 89129-110
DIGE Buffer Kit, Cytiva
Buffers Electrophoresis Buffers Running Buffers
DIGE buffer kit is a tool to increase the quality, reproducibility, and convenience of 2-D DIGE using the Ettan™ DIGE system. The DIGE buffer kit is a specially designed two-buffer system for running up to twelve DIGE gels simultaneously.
  • Precast 12,5% polyacrylamide buffer for second-dimension 2-D electrophoresis
  • Ready-to-use products with a long shelf-life that saves you time by eliminating the need to make buffers
  • DIGE gels run with DIGE buffer kit result in spot maps with higher reproducibility compared to homecast polyacrylamide gels
DIGE gel should be used with the DIGE buffer kit, which consists of concentrated running buffers and sealing solution for attaching immobilinedrystrip gels (IPG strips) to the top of the polyacrylamide gel. The capacity of the buffer system is similar to the used laemmli (tris-glycine) buffer system, and the separation performance of DIGE gel is comparable to 12.5% laemmli gels (1). DIGE gel and DIGE buffer kit offer the high quality and performance of the laemmli system with the added advantages of convenience and time savings. DIGE gel is intended for use with the DIGE buffer kit and ettan dalt large vertical system. ettan dalttwelve and ettan daltsix instruments are designed for large-format, 2-D electrophoresis using either 18 or 24 cm immobiline drystrip gels. with ettan dalttwelve, up to 12 DIGE gels can be run simultaneously with separations complete in 4 to 6 hours. The DIGE buffer kit contains the amount of reagents necessary for a single run of up to 12 DIGE gels using the ettan dalttwelve electrophoresis unit, or two runs of up to six gels using the ettan daltsix unit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty