Would you like to log in again?
BINDER STATIC DISSIP 2IN BLUE
Supplier: DESCO
Static-Dissipative Vinyl Binders^ Static-dissipative vinyl stretched over cardboard. Three-ring binders. Can be used in ESD-sensitive environments. Edges are heat-sealed. Hold standard 21.6 x 27.9cm paper. Color: Light blue. Ring size: 5.08cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BINDER STATIC DISSIP 2IN BLUE 89129-440 07415 Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty