Would you like to log in again?
DuPont™ Tyvek® Micro-Clean® 2-1-2 Coveralls
Supplier: DuPont
Tyvek® Micro-Clean®
CC252BBU3X0025PI CC252BBU2X00250S CC252BBULG0025PI CC252BBUSM00250S CC252BBUSM0025PI CC252BBUXL00250S CC252BBU2X00250S CC252BBU2X0025PI CC252BBUMD00250S CC252BBULG00250S CC252BBU2X0025PI CC252BBUMD00250S CC252BBU3X00250S CC252BBUXL00250S CC252BBUSM0025PI CC252BBU5X00250S CC252BBU4X00250S CC252BBU3X0025PI CC252BBU3X00250S CC252BBUMD0025PI CC252BBUXL0025PI CC252BBULG0025PI CC252BBU4X0025PI CC252BBUMD0025PI CC252BBU4X0025PI CC252BBUSM00250S CC252BBU4X00250S CC252BBULG00250S CC252BBU5X00250S CC252BBUXL0025PI
89127-156CS 805.41 USD InStock
CA89127-144 CA89127-166 89127-156 CA89127-164 89127-154 CA89127-148 CA89127-146 89127-158 89129-588 CA89127-162 CA89129-592 89127-152 CA89127-160 89127-150 CA89129-590 CA89129-588 89127-146 CA89127-154 89129-590 89127-144 89127-166 CA89127-152 CA89127-158 89127-148 CA89127-156 89127-160 89127-164 CA89127-150 89127-162 89129-592
DuPont™ Tyvek® Micro-Clean® 2-1-2 Coveralls
Disposable Garments
Tyvek® Micro-Clean® 2-1-2 delivers the particle barrier, durability and comfort of Tyvek® with a unique blue color.
  • Bound seams
  • Bound neck with loop at center back
  • Dolman sleeve design
  • Covered elastic wrists and ankles
  • Zipper closure
Proprietary blue polymeric resin coating is applied to Tyvek® fabric. Dolman sleeve design allows for greater range of motion and comfort. Coveralls have elastic openings for tighter fit at wrists and ankles. Covered elastic minimizes particle shedding. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration.
Certifications: Sterile garments are gamma sterilized to a Sterility Assurance Level (SAL) of 10-6.
Packaging: Coveralls are individually packaged in an opaque bag.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty