Would you like to log in again?
DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves and Snaps
Supplier: DuPont
Tyvek® IsoClean®
IC254BWHMD0025CS IC254BWHLG0025CS IC254BWHXL0025CS IC254BWH4X0025CS IC254BWHXL0025CS IC254BWH2X0025CS IC254BWHMD0025CS IC254BWH3X0025CS IC254BWH3X0025CS IC254BWH2X0025CS IC254BWHSM0025CS IC254BWH4X0025CS IC254BWHLG0025CS IC254BWHSM0025CS
89129-664CS 905.21 USD InStock
CA89129-674 89129-664 CA89129-662 89129-674 CA89129-672 89129-662 CA89129-670 89129-672 89129-670 CA89129-668 CA89129-666 89129-668 89129-666 CA89129-664
DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves and Snaps
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound seams
  • Bound neck
  • Dolman sleeve design
  • Elastic wrists and ankles
  • Zipper closure
  • Snaps for aseptic donning
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coveralls feature elastic openings at wrists and ankles for a tighter fit. Bound neck allows for lower particle shedding. Snaps at sleeves and legs to aid in aseptic donning. Front zipper closure allows for easy donning and doffing. Dolman sleeve design allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty