Would you like to log in again?
100% Polyester Sealed Edge Cleanroom Wiper, Berkshire
Supplier: Berkshire
CPSLP1212.14 CPSLP.0909.8
89129-854CS 1383.53 USD InStock
89129-854 89129-850
100% Polyester Sealed Edge Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
CapSure®-LP features an innovative process, that enables wipers to capture and retain more contaminant particles to provide a more effective wipe and reduce the risk of recontamination.
100% continuous filament polyester knit with sealed edges captures 42× more particles than competitive wipers and retains 93% of particles initially captured. Minimal particles, fibers, ions and extractables. Airborne particle release 80% lower than competitive wipers. Designed for use in ISO class 3 and higher cleanroom environments.
Applications include oxidation, metallization, CVD or photolithography processes. Chamber cleaning and CMP processing, stencil and other print roll cleaning applications, cleaning of medical device products, applying and removing cleaning and disinfecting solutions.
Ordering information: Quality control and lot traceability identification on each case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty