Would you like to log in again?
Prostate Biopsy Kits, Azer Scientific
Supplier: Azer Scientific
10-831 10-820 10-816 10-816
89129-904CS 327.25 USD InStock
89129-904 89129-902 CA89129-906 89129-906
Prostate Biopsy Kits, Azer Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
These ready-to-use kits are packaged with 8, 12, or 16 vials, which are prefilled with 20 ml of 10% neutral buffered formalin and a leak-resistant seal.
  • Cardboard insert labeled with the appropriate section of the prostate
Each container is half-filled with 10% neutral buffered formalin.
The insert of each kit is labeled with the appropriate section of the prostate, which allows for easy tissue sampling.
Certifications: Vials are 95 kPA compliant.
Ordering information: Kits are also available with components, including a biohazard bag, formalin absorbent pad, and desiccant pack.
Packaging: Eight 20 ml Formalock™ containers are included. Vials are prefilled with 20 ml of 10% neutral buffered formalin and a leak-resistant seal. Each case contains 25 kits.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty