Would you like to log in again?
jetPEI®, HTS DNA Transfection Reagent
Supplier: Polyplus-transfection
jetPEI®
101000020 101000053 101000020 101000053
89129-918EA 1888.38 USD InStock
CA89129-918 89129-918 CA89129-916 89129-916
jetPEI®, HTS DNA Transfection Reagent
Transfection Reagents
jetPEI® transfection reagent is a linear polyethylenimine derivative, free of components of animal origin, providing highly effective and reproducible gene delivery, perfectly suited for High Throughput Screening (HTS) applications.
  • Well-suited for adherent and suspension cells
  • Fast and efficient methods to transfect cells for HTS
  • Compatible with serum and antibiotics
  • Exceptionally reproducible results
  • Reverse, batch & forward protocols available
jetPEI® transfection reagent is particularly well suited for automated or manual High Throughput Screening (HTS) with three protocols available: Reverse, batch and forward.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of jetPEI® is tested in-house by DNA transfection of HeLa cells. Transfection with a firefly Luciferase gene under the control of CMV promoter gives at least 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Delivery information: 150 mM NaCl solution is included. 1 mL of jetPEI® transfection reagent is sufficient to perform 2000 transfections in 96-well plates
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty