Would you like to log in again?
Accessories for Gram Stain Kits, Azer Scientific
Supplier: Azer Scientific
ES806 ES802 ES812-2.5G ES806 ES802 ES808 ES812-2.5G
89130-196EA 345.04 USD InStock
CA89130-192 CA89130-194 89130-196 89130-190 89130-192 CA89130-190 CA89130-196
Accessories for Gram Stain Kits, Azer Scientific
Stains Histology Stains
These gram stain kits are used to determine the Gram reaction for microorganism identification.
  • Convenient Spout Caps
  • Comprehensive Product Insert
  • One Bottle of Each: Stabilized or Unstabilized Iodine, Crystal Violet, Decolorizer, and Safranin
Kits contain one 237mL (8oz.) bottle of each component: stabilized or unstabilized iodine, crystal violet, decolorizer, and safranin.
Detailed product insert and convenient spout caps are also included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty