Would you like to log in again?
96-Well Non-binding Microplates, Non-Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
655906 655904 651901 655903 655900 650901 655901
89131-678CS 358.61 USD InStock
89131-678 89131-680 89131-672 89089-582 89130-330 89131-676 89131-674
96-Well Non-binding Microplates, Non-Sterile, Greiner Bio-One
Microplates HTS Microplates
Greiner Bio-One's non-binding microplate surfaces prevent unwanted non-specific binding.
  • Non-binding
  • Nonsterile
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
The surface is characterised by low protein, DNA and peptide binding properties to significantly improve data quality, especially in biochemical assays
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
Low amounts of biomolecular binding can cause an undesirable increase in background, resulting in decreased signal-to-noise ratio. The non-binding microplate surfaces significantly increase assay sensitivity by reducing background and improving signal-to-noise ratio. Non-binding microplates are available in black, white, and clear. Microplates include solid and µClear film well bottoms.
Ordering information: Lid not included
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty