Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Line Valve, Ball, Stainless Steel, Rotarex
Supplier: Rotarex
4SBV2TSS
89130-356EA 327.05 USD InStock
89130-356
Line Valve, Ball, Stainless Steel, Rotarex
Valves Line Valves
Used for instrumentation, hydraulic pipes, pneumatic pipes, chromatography, control panels, moderate, vacuum systems, chemical research, gas analysis, etc.
Diaphragm sealing.
The assembly is made quickly by inserting the tube in the ready-to-use fitting and tighten. They require no special tools.
Ordering information: Additional materials of construction and end connections are available available; contact your VWR sales representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty