Would you like to log in again?
Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Supplier: Rotarex
SL1544
89130-368EA 1192.54 USD InStock
89130-368
Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Gas Regulators
Used as a line regulator for high-flow industrial or lab applications.
As a second level of regulation the S 15 will supply a precise outlet pressure to the process. Can be used for wall or panel mounting with its compact design, the rear threads and fixing ring.
Pressure control with Balanced-Valve Technology: the effect of inlet pressure fluctuations on outlet pressure are minimized. BV-technology delivers an exceptionally stable outlet pressure and flow even with high flow line regulators. Longer useful regulator lifetime and lower total ownership cost.
Ordering information: Additional outlet pressures and flow rates available; contact your Avantor sales representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty