Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NucliSENS® easyMAG® Reagents and Accessories for Automated Extraction of RNA/DNA, bioMerieux
Supplier: bioMérieux
280134 280131 280132 280146 280130 200293 280133 200292
89130-518EA 2413.91 USD InStock
89130-518 89130-526 89130-528 89130-522 89130-533 89130-524 89130-534 89130-530
NucliSENS® easyMAG® Reagents and Accessories for Automated Extraction of RNA/DNA, bioMerieux
Automated Nucleic Acid Purification Systems
The NucliSENS® easyMAG® accessory products are reagents and disposables to be used with the bioMérieux instruments which mention these products in their User Manuals
  • Disposables
  • For bioMerieux instruments
  • Enable automated extraction
They enable the automated extraction (purification and concentration) of total nucleic acids (RNA/DNA) from homogeneous human biological fluid samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty