Would you like to log in again?
Courier Specimen Lock Boxes, TAGG Industries
Supplier: TAGG Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
50114 50113 50111 50118 50112
89130-878CS 371.47 USD InStock
89130-878 89130-874 89130-876 89130-880 89130-872
Courier Specimen Lock Boxes, TAGG Industries
Security Locks
Specimen Courier Lock Boxes are designed to provide a secure option for after-hours pick-up or delivery
  • All boxes are supplied with two keys
  • Boxes are designed to hang from the top of any office door
  • Under door strap is available for the Plastic/Biocare Lock Box
  • Traditional Lock Box and Plastic/Biocare Lock Box feature foam lining insulation
  • Keyless access by client available on Dual Door Lock Boxes
  • Dual Door Lock Boxes feature foam inserts for tube storage
  • X-Large Traditional Lock Box is designed to be placed on the floor, rather than hung from a door
All boxes feature durable material construction, with either metal or injection molded ABS plastic options available.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty