Would you like to log in again?
VWR® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged
Supplier: VWR
VWR®
4037 4042A 4044 4031A 4043 4032 4033 4038 4045 4035A 4030A 4034 4047 4039 4046 4036A 4040 4041A
76097-926CS 377.03 USD InStock
76097-926 76097-924 89130-890 76097-922 89130-908 89130-914 76093-886 89130-916 89130-900 76093-884 89130-910 89130-888 76093-882 89130-902 89130-912 89130-896 89130-898 89130-886
VWR® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged
Pipettes Serological Pipettes
VWR® Disposable Serological Pipettes are manufactured from polystyrene and are supplied sterile and plugged.
  • Calibrated to deliver
  • Color-coded for quick identification
  • Equipped with ascending, descending, and negative graduations
Polystyrene construction for sterile serological and tissue culture applications. Easy-to-read black ascending and descending graduations simplify fluid measuring and dispensing. All pipettes feature negative graduations for extra working volume, and are color-coded for quick size identification. Accuracy is ±2% at full volume.
Certifications: All pipettes are in compliance with ASTM Standard E-934, Standard Specification for Serological Pipettes, Disposable Plastic. DNase- and RNase-free.
Packaging: Available in three packaging options: Individually wrapped in clear, easily recyclable, all-plastic polyethylene with pop-through opening for aseptic removal; individually wrapped in color-coded paper/plastic that reduces static cling; or bulk packed in polyethylene bags for high volume liquid handling applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty