Would you like to log in again?
Zinc Chelating Resin for Immobilized Metal Affinity Chromatography Purification, G-Biosciences
Supplier: G-Biosciences
786-287 786-287
89131-012EA 183.62 USD InStock
89131-012 CA89131-012
Zinc Chelating Resin for Immobilized Metal Affinity Chromatography Purification, G-Biosciences
Protein Purification Protein Purification Resins
G-Biosciences' Zinc Chelating Resin is an immobilized metal affinity chromatography (IMAC) resin that was specifically designed to utilize Zn²⁺ for the purification of zinc binding proteins, including 6x histidine tagged proteins.
G-Biosciences' Zinc Chelating Resin binds to six histidine residues (6x His), a common tag used in protein purification. The resin consists of iminodiacetate coupled to 6% cross-linked agarose beads. The iminodiacetate binds divalent zinc ion with a capacity of 20-40μmoles Znu²⁺/mL resin. The protein binding capacity is >20 mg protein per mL resin. We have demonstrated binding of >40 mg of a 50 kDa 6X His tagged proteins to a mL of resin.
G-Biosciences' Immobilized Nickel, Cobalt and Copper Chelating Resins are also available. Cobalt has the highest selectivity followed by Zinc, Nickel then Copper, but has the lowest loading capacity. Copper has the highest loading capacity, followed by Nickel then Zinc. The capacity of zinc is >20 mg per mL. Specific binding/wash and elution buffers are available.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty