Would you like to log in again?
Statshield® Metal-In Bags, Desco Industries
Supplier: Desco Industries
Statshield®
13430 13415 13420 13725 13665 13610 13720 13506 13460 13475 13675 13515
89131-070PK 30.88 USD InStock
89131-070 47731-604 89131-042 89131-052 89131-040 11000-488 89131-050 89131-072 89131-068 89131-056 89131-054 47731-519
Statshield® Metal-In Bags, Desco Industries
Bags Storage Bags
Metal-in ESD shielding bag with low charging interior and exterior layers.
  • Identified with ESD protective symbol
  • Polycarbonate compatible containing no amines or N-octanic acid
  • Testable to industry standards
  • Low charging antistatic high visibility yellow reclosable zipper
  • Made in The United States of America
Most commonly used shielding bag meets limits of ANSI/ESD S541 per ANSI/ESD S20.20. 40% light transmission allows for easy identification of bag contents. Integral antistatic, low tribocharging properties mean contents will not electrostatically charge during movement. Integral static dissipative properties are not topically treated, not humidity dependant, and won't wear off. Manufactured on specially designed soft fold converting machines to prevent scratches and creases, maintaining the integrity of Faraday Cage shielding protection. Large ³/₈" wide seals provide greater seal strength compared to comparable products with ¹/₈" wide seals.
Certifications: Meets limits of ANSI/ESD S541 per ANSI/ESD S20.20. Lead-free, RoHS compliant.
Packaging: Statshield® bags are packaged 100 per package in an oversized shielding bag rather than a cardboard box. The bags are not exposed to water vapors that will degrade the metallized shielding layer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty