Would you like to log in again?
Humidity Indicator Card, Desco
Supplier: Desco Industries
13870
89131-360EA 37.51 USD InStock
89131-360
Humidity Indicator Card, Desco
Test Paper Indicator Paper
Reversible Humidity Indicator Cards for vapor protection measure the level of humidity that the contents in package are exposed.
  • For use with Statshield® ESD Shielding Moisture Barrier Bags
  • Humidity indicator spots are reversible
  • 100 cards per can
  • Made in the United States of America
Cards measure the level of humidity that the contents of the packaging have been exposed to. They can be used with Statshield® ESD Shielding Moisture Barrier Bags to provide protection for moisture sensitive items such as surface mount devices. Reversible design allows the card to be reused, providing better value.
Certifications: Meets MIL-I-8835A And JEDEC J-STD-033D (Type 1) Table 3-1 Dry Packing requirements. Lead-Free, RoHS compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty