Would you like to log in again?
Advanced TC™ Treated Cell Culture Multiwell Plate with Lid, Polystyrene, Greiner Bio-One
Supplier: Greiner bio-one
662960 677980 657960 665980
89131-690CS 438.03 USD InStock
89131-690 89136-866 89131-688 89136-864
Advanced TC™ Treated Cell Culture Multiwell Plate with Lid, Polystyrene, Greiner Bio-One
Microplates Cell Culture Plates
Manufactured from optically clear virgin polystyrene, these flat-bottom plates are supplied sterile and pyrogen-free with a printed lot number for traceability.
  • Clear virgin polystyrene construction
  • High degree of optical clarity
  • Nonpyrogenic
  • Reduced risk of cross-contamination
  • Uniform design for manual or automated handling
The chimney well design and low evaporation lids with condensation rings reduce the risk of cross-contamination
Advanced Tissue Culture (TC) treated Cell Culture Multiwell Plates allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions. Advanced TC surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield. Clear Polystyrene lid with condensation rings. All plates come with Lid. Advanced TC Treated for all plates.
Sterilized by radiation, DNase- Free, RNase- Free, human DNA - Free, nonpyrogenic and noncytotoxic.
Plates are available tissue culture treated for the culture of anchorage-dependent cells or nontreated for suspension cell culture applications. Excellent heat exchange within each plate is ensured during incubation through air circulation spaces between stacked plates. Uniform design makes them suitable for manual and automated handling. The well depth and diameter also allows the use of specific tissue culture inserts.
Ordering information: Sterile and user friendly packaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty