Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Supplier: Labplas
Twirl'Em®
EFL47012E EPR-4590E EPR-7012E EPL-7015E EPR-3070E EPL-5590E EFR-3070E EPL-4590E EPL-3070E EFR47012E EFL-5512E EFR-4590E EFL-1012E EFR-5590E EPL-5515E EFL-4590E EPR45590E EPR-3050E EFR-7012E EPR45590E EPL-7012E EPL-5515E EFR-1015E EFL-7012E EFL47012E EFL-1012E EPR-7012E EPR-559E-VW1 EPR-5590E EFL-5590E EFR-3070E EPL-4590E EPL47012E EPL-7012E EFR-4590E EFR-1012E EFL-4590E EFL-5512E EFR47012E EPR47012E EFL-5590E EPL-7015E EFR-7012E EPR-4590E EPL45590E EPR47012E EFR-1012E EFL-1015E EPL47012E EPL45590E EPL-5590E
89131-834CS 744.17 USD InStock
CA89131-802 89131-834 CA89131-804 89131-832 CA89131-800 89131-794 89131-792 89131-830 89131-790 89131-824 CA89131-834 CA89131-836 89131-788 CA89131-830 CA89131-798 CA89131-832 89131-826 CA89131-794 89131-782 CA89131-796 89131-820 CA89131-790 89131-786 89131-784 CA89131-792 89131-818 CA89131-828 CA89131-824 89131-812 CA89131-826 89131-810 CA89131-786 CA89131-820 CA89131-788 89131-814 CA89131-822 CA89131-782 CA89131-784 CA89131-780 89131-808 CA89131-816 CA89131-818 89131-802 CA89131-812 89131-800 CA89131-814 89131-806 CA89131-810 89131-804 CA89131-806 CA89131-808
TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Bags Sample Bags
These biodegradable sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals
  • Feature a wire closure for an airtight seal
  • Wide-sealed bottoms for added leakproof protection
They are easy to use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal. Round wire bags have two wires for closure and puncture-proof safety tabs to eliminate sharp points that can cause bag puncture and abrasion. Bags with white marking area allow for written notation.
Certifications: Bags comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty