Would you like to log in again?
CleanTube™ Tube Wipes, Contec
Supplier: Contec
LWTW0032
89131-840CS 177.42 USD InStock
89131-840
CleanTube™ Tube Wipes, Contec
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The CleanTube™ tubular wipe from Contec brings the cleanliness of ISO Class 4 cleanroom laundering to other critical manufacturing industries.
  • Ideal for removing excess sealant during automotive or aerospace assembly
  • Excellent on coarse or rough sanded surfaces
  • Ideal for equipment cleaning and maintenance
Durable polyester provides excellent sorbency. Available in a variety of textures for superior cleaning and pick-up performance. Laundered in an ISO Class 4 cleanroom. Heat cauterized edges naturally roll outward, reducing the potential for particles from cut fibers. The tubular construction of CleanTube™ wipes allows technicians to "load" their arms with several wipes at a time, reducing downtime. High stretch allows for cleaning in tight spaces. Economically bulk packaged. Multiple packaging configurations are available.
Constructed of 100% polyester knit and available in a variety of lengths and textures, CleanTube™ wipes are ideal for many surface preparation applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty