Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
MAPM02C68 MAP003C36 MAP100C37 MAP100C37 MAP001C37 MAP030C38 MAP030C38 MAP001C36 MAP003C38 MAPM45C68 MAP100C36 MAP010C38 MAPM02C68 MAP003C37 MAPM45C68 MAPM02C67 MAP100C38 MAP010C38 MAP003C37 MAP010C36 MAP001C38 MAPM02C67 MAP010C37 MAP003C38 MAP001C38 MAP001C36 MAPM45C67 MAP001C37 MAP030C36 MAP010C36 MAP003C36 MAP100C38 MAP030C37 MAP010C37 MAPM45C67 MAP030C37 MAP030C36 MAP100C36
89131-978PK 78.85 USD InStock
CA89131-988 89131-978 89131-998 89131-976 CA89233-884 CA89131-984 CA89233-882 CA89131-986 89131-992 CA89233-880 CA89131-980 89131-990 CA89131-982 89131-974 89131-996 89131-972 89131-994 89132-000 89132-002 89131-988 CA89131-978 89131-986 CA89131-974 CA89131-996 89233-884 CA89131-976 CA89131-998 CA89131-992 89131-980 89233-880 89233-882 CA89131-994 CA89131-972 89131-984 89131-982 CA89131-990 CA89132-002 CA89132-000
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Filters Centrifugal Filters
These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.
  • Provides high recoveries, typically >90%
  • Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding
  • Versatile ultrafiltration Omega™ membrane is available in a variety of MWCOs
  • Built-in deadstop prevents spinning to dryness
  • Color-coded and laser etched for easy identification
Macrosep™ Advance centrifugal devices can be used for a wide range of ultrafiltration applications including the concentration and desalting of proteins, buffer exchange and protein fractionation.
The design of the devices maximizes the effective filtration area allowing for rapid sample processing while maintaining a gentle concentration environment to preserve protein activity and conformation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty