Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
MCP030C46 MCPM02C68 MCP001C41 MCP030C41 MCP001C46 MCPM02C68 MCP030C46 MCPM02C67 MCP001C46 MCPM45C68 MCPM02C67 MCP010C46 MCPM45C67 MCP003C46 MCP100C46 MCP010C41 MCP030C41 MCP010C46 MCP003C41 MCP100C41 MCP100C41 MCP100C46 MCPM45C67 MCP003C46 MCP003C41 MCP010C41 MCP001C41
89233-878PK 578.55 USD InStock
89233-878 89233-876 89132-006 CA89132-018 89132-008 CA89132-016 89132-020 CA89132-010 89132-022 CA89132-014 89132-024 CA89132-012 89132-026 89132-004 CA89233-878 CA89233-876 89132-018 CA89132-006 CA89132-004 CA89132-008 CA89132-020 89132-010 89132-012 89132-014 CA89132-024 CA89132-022 89132-016
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Filters Centrifugal Filters
Precise, quick recovery of microliter volumes of concentrate from starting volumes up to 5.0 ml.
  • Available with low protein-binding OmegaTM and Supor® (PES) membranes
  • Omega modified PES membranes come in a wide range of MWCOs, color-coded for easy identification
  • With Omega membranes you can Achieve 50X concentration and >90% recovery in just minutes
  • Supor (PES) membranes come in 0.45 and 0.2 µm pore sizes for microfiltration applications
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent bypass or seal failure
For smaller samples use Pall Nanosep ®centrifuge filters. For larger samples use Pall Macrosep® or Jumbosep™ centrifugal filter.
Microsep® Centrifugal devices with Omega (modified polyethersulfone) membrane for ultrafiltration are ideal for concentration of dilute protein samples, buffer provides an efficient way to remove or exchange salts, as well as fractionating molecules of different molecular weights. Supor (PES) microfiltration devices are ideal for clarifying samples with gross particulate, as well as removing proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies.
A built-in deadstop feature prevents samples from spinning to dryness and the unique paddle shape design prevents the creation of a density gradient, preventing proteins from aggregating. Filters are color coded and laser etched for easy identification. The Microsep sample reservoir and filtrate receiver are constructed of polypropylene to reduce non-specific binding of biomolecules.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty