Would you like to log in again?
Pig Blue® Absorbent Mat Pad, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BLU101 BLU100
89132-768CS 168.86 USD InStock
89132-768 89132-764
Pig Blue® Absorbent Mat Pad, New Pig
Spill Control Materials
Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention. FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent. Stays together, even when picking up or shifting a fully saturated mat.
  • Absorbs liquid evenly for better wicking and retention
  • Super-absorbent natural fibers with strong synthetic fibers for high-performance absorbent
  • Stays together, even when picking up or shifting fully saturated mat
Contains 75% recycled fibers; ideal in situations where green products are desired or required
Heavyweight Pig Blue construction is super absorbent for your most demanding tasks and large-volume cleanups. Lightweight Pig Blue construction is ideal for your everyday tasks and cleanups.
Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08. Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty