Would you like to log in again?
MetriZyme™ Dual Enzymatic Instrument Detergent, Metrex
Supplier: Metrex
MetriZyme™
10-4000
89132-869CS 623.29 USD InStock
89132-869
MetriZyme™ Dual Enzymatic Instrument Detergent, Metrex
Cleaning Agents Detergents and Surface Cleansers
MetriZyme™ is designed for superior performance in the presence of high organic debris while offering the broadest cleaning action on a variety of protein soils
  • Powerful cleaning
  • Ready to use
  • Liquid concentrate
Dual enzymatic instrument detergent includes an enzyme stabilizer, a buffer to maintain pH, a preservative, an antifoaming agent and solution dye to differentiate it from water and other solutions.
Surfactants provide additional cleaning on carbohydrates, lipids and protein soils.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty