Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Columbia Blood Agar Base Dehydrated Culture Media, HiMedia

Supplier: HiMedia Laboratories
M144-2.5KG M144-500G M144-2.5KG M144-5KG M144-5KG M144-500G
89132-898EA 768.44 USD InStock
89132-898 CA89132-898 95021-134 CA95021-134 CA89224-036 89224-036
Columbia Blood Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
An efficient base for the preparation of blood agar or chocolate agar, and for various selective and identification media and isolation of organisms from clinical and non clinical samples.

  • Cream to yellow homogeneous free flowing powder
  • Firm gelling, comparable with 1.5% Agar gel
  • pH: 7.10 to 7.50
  • Store below 10 to 30 °C

These agar bases contain peptone, special (23.000 g/l); corn starch (1.000 g/l); sodium chloride (5.000 g/l) and agar (15.000 g/l).

Certifications: In accordance with the harmonazied method of USP/EP/BP/JP.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty