Would you like to log in again?
Agarose, Pulse Field Gel Electrophoresis, IBI Scientific
Supplier: IBI Scientific
IB70061 IB70064 IB70065 IB70060 IB70063 IB70062
89133-114EA 249.91 USD InStock
89133-114 89133-112 89133-110 89133-108 89133-118 89133-116
Agarose, Pulse Field Gel Electrophoresis, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
Pulse Field Gel Electrophoresis Agarose is a linear polymer with a high molecular weight, creating gel structures unlike those of standard agarose
Some features of IBI PFGE Agarose are extrememly high gel strength, greater thermal stability when compared to standard agarose, and exceptionally low absorption of staining reagents.
IBI PFGE Agarose can be used for pulse field gel electrophoresis, conventional electrophoresis and blotting, and cell and enzyme immobilization. PFGE Agarose will separate and finely resolve molecules > 1000bp.
This characteristic, along with its very low sulfate content produces a very strong intercatenary interaction, resulting in a gel with a very high gel strength and high exclusion limit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty