Would you like to log in again?
SCRUBWRIGHT BELT DIA 280 GRIT
Supplier: FOAMTEC INTERNATIONAL, LLC.
Diamond ScrubBELTS 280 grit^ .25 x 12.75in belt Diamond 280 grit for use with the ScrubWRIGHT PEN. Creates a versatile instrument perfect for polishing stains, scratches and process-induced residue from scratch sensitive surfaces. 5 belts per pack, 5 packs per case.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SCRUBWRIGHT BELT DIA 280 GRIT 89133-228 HT4528DW-5 Case of 25 Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty